COM448686095 Combi-Blocks, Inc.

methyl (2R)-2-acetamido-4-(methylsulfanyl)butanoate

  • MDL number MFCD02094377
  • Cas Number : 114285-15-3
  • Molecular Weight : 205.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0528

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0528
Availability details for
Quantity
  • Availability
  • Add to Cart