COM448686098 Combi-Blocks, Inc.

Methyl (2S,3aS,7aS)-octahydro-1H-indole-2-carboxylate

  • MDL number MFCD20662144
  • Cas Number : 192436-84-3
  • Molecular Weight : 183.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 96
Storage temp. Refrigerated

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0531
Availability details for
Quantity
  • Availability
  • Add to Cart