COM448686099 Combi-Blocks, Inc.

methyl (2R)-2-{[(tert-butoxy)carbonyl](methyl)amino}-4-methylpentanoate

  • MDL number MFCD27578242
  • Cas Number : 1820572-12-0
  • Molecular Weight : 259.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0532

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0532
Availability details for
Quantity
  • Availability
  • Add to Cart