COM448686100 Combi-Blocks, Inc.

1-tert-butyl 4-methyl (2R)-2-{[(tert-butoxy)carbonyl]amino}butanedioate

  • MDL number MFCD27578243
  • Cas Number : 77004-76-3
  • Molecular Weight : 303.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0533

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0533
Availability details for
Quantity
  • Availability
  • Add to Cart