COM448686105 Combi-Blocks, Inc.

3,5-Bis(trifluoromethyl)cinnamic acid methyl ester

  • MDL number MFCD01861885
  • Cas Number : 705250-77-7
  • Molecular Weight : 298.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0539

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0539
Availability details for
Quantity
  • Availability
  • Add to Cart