COM448686107 Combi-Blocks, Inc.

Methyl 2-(2,2,2-trifluoroacetamido)acetate

  • MDL number MFCD00056333
  • Cas Number : 383-72-2
  • Molecular Weight : 185.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0546

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0546
Availability details for
Quantity
  • Availability
  • Add to Cart