COM448686109 Combi-Blocks, Inc.

methyl 2-(acetyloxy)-2-phenylacetate

  • MDL number MFCD27578245
  • Cas Number : 947-94-4
  • Molecular Weight : 208.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0548

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0548
Availability details for
Quantity
  • Availability
  • Add to Cart