COM448686111 Combi-Blocks, Inc.

Methyl 2,3-dimethoxy-5-sulphamoylbenzoate

  • MDL number MFCD01317563
  • Cas Number : 66644-82-4
  • Molecular Weight : 275.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0550

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0550
Availability details for
Quantity
  • Availability
  • Add to Cart