COM448686114 Combi-Blocks, Inc.

D-Arginine methyl ester hydrochloride

  • MDL number MFCD18070884
  • Cas Number : 111237-99-1
  • Molecular Weight : 224.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0557

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0557
Availability details for
Quantity
  • Availability
  • Add to Cart