COM448686115 Combi-Blocks, Inc.

methyl (2S)-2-(phenylformamido)pentanoate

  • MDL number MFCD27578246
  • Cas Number : 1492-14-4
  • Molecular Weight : 235.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0561

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0561
Availability details for
Quantity
  • Availability
  • Add to Cart