COM448686116 Combi-Blocks, Inc.

Methyl (2S)-2-amino-3-(3,5-difluorophenyl)propanoate

  • MDL number MFCD08058345
  • Cas Number : 473567-59-8
  • Molecular Weight : 215.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0562

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0562
Availability details for
Quantity
  • Availability
  • Add to Cart