COM448686117 Combi-Blocks, Inc.

methyl (2S)-2-{[(tert-butoxy)carbonyl]amino}-4-methanesulfonylbutanoate

  • MDL number MFCD27578247
  • Cas Number : 160141-86-6
  • Molecular Weight : 295.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0564

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0564
Availability details for
Quantity
  • Availability
  • Add to Cart