COM448686124 Combi-Blocks, Inc.

4-Propoxy-benzoic acid methyl ester

  • MDL number MFCD00084854
  • Cas Number : 115478-59-6
  • Molecular Weight : 194.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0576

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0576
Availability details for
Quantity
  • Availability
  • Add to Cart