COM448686126 Combi-Blocks, Inc.

methyl (2R)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}pentanoate

  • MDL number MFCD27578256
  • Cas Number : 1820569-85-4
  • Molecular Weight : 353.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0579

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0579
Availability details for
Quantity
  • Availability
  • Add to Cart