COM448686131 Combi-Blocks, Inc.

methyl (2S,3R)-3-(tert-butoxy)-2-{[(tert-butoxy)carbonyl]amino}butanoate

  • MDL number MFCD27578259
  • Cas Number : 1262482-76-7
  • Molecular Weight : 289.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0584

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0584
Availability details for
Quantity
  • Availability
  • Add to Cart