COM448686133 Combi-Blocks, Inc.

methyl 2-ethoxypyridine-4-carboxylate

  • MDL number MFCD27578260
  • Cas Number : 153928-57-5
  • Molecular Weight : 181.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0589

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0589
Availability details for
Quantity
  • Availability
  • Add to Cart