COM448686142 Combi-Blocks, Inc.

D-Cysteine methyl ester

  • MDL number MFCD19203230
  • Cas Number : 88806-98-8
  • Molecular Weight : 135.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0601

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0601
Availability details for
Quantity
  • Availability
  • Add to Cart