COM448686145 Combi-Blocks, Inc.

methyl (2S)-2-amino-4-phenylbutanoate hydrochloride

  • MDL number MFCD27578263
  • Cas Number : 60425-49-2
  • Molecular Weight : 229.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0606

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0606
Availability details for
Quantity
  • Availability
  • Add to Cart