COM448686146 Combi-Blocks, Inc.

methyl (2S)-1-[(2S)-2-methyl-3-sulfanylpropanoyl]pyrrolidine-2-carboxylate

  • MDL number MFCD27578264
  • Cas Number : 176036-41-2
  • Molecular Weight : 231.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0607

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0607
Availability details for
Quantity
  • Availability
  • Add to Cart