COM448686148 Combi-Blocks, Inc.

9-Anthracenecarboxylic acid methyl ester

  • MDL number MFCD00095211
  • Cas Number : 1504-39-8
  • Molecular Weight : 236.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0614

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0614
Availability details for
Quantity
  • Availability
  • Add to Cart