COM448686151 Combi-Blocks, Inc.

methyl (2S)-2-{[(tert-butoxy)carbonyl]amino}-3-[(triphenylmethyl)carbamoyl]propanoate

  • MDL number MFCD27578265
  • Cas Number : 161001-61-2
  • Molecular Weight : 488.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0618

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0618
Availability details for
Quantity
  • Availability
  • Add to Cart