COM448686155 Combi-Blocks, Inc.

methyl (2S)-2,5-bis({[(tert-butoxy)carbonyl]amino})pentanoate

  • MDL number MFCD27578268
  • Cas Number : 97347-27-8
  • Molecular Weight : 346.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0624

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0624
Availability details for
Quantity
  • Availability
  • Add to Cart