COM448686159 Combi-Blocks, Inc.

3-Methyl-4-nitrophenylacetic acid methyl ester

  • MDL number MFCD18399608
  • Cas Number : 156480-27-2
  • Molecular Weight : 209.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0630

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0630
Availability details for
Quantity
  • Availability
  • Add to Cart