COM448686164 Combi-Blocks, Inc.

methyl 1-cyclopropyl-6-fluoro-4-oxo-7-(piperazin-1-yl)quinoline-3-carboxylate

  • MDL number MFCD27578272
  • Cas Number : 107884-23-1
  • Molecular Weight : 345.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0637

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0637
Availability details for
Quantity
  • Availability
  • Add to Cart