COM448686166 Combi-Blocks, Inc.

2,4,6-Trimethyl-benzeneacetic acid methyl ester

  • MDL number MFCD12024951
  • Cas Number : 41841-19-4
  • Molecular Weight : 192.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0639

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0639
Availability details for
Quantity
  • Availability
  • Add to Cart