COM448686168 Combi-Blocks, Inc.

methyl (2S)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-2-phenylacetate

  • MDL number MFCD27578274
  • Cas Number : 1820574-62-6
  • Molecular Weight : 387.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0644

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0644
Availability details for
Quantity
  • Availability
  • Add to Cart