COM448686169 Combi-Blocks, Inc.

Methyl (2e)-3-(4-ethoxyphenyl)prop-2-enoate

  • MDL number MFCD09755017
  • Cas Number : 77891-34-0
  • Molecular Weight : 206.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0646

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0646
Availability details for
Quantity
  • Availability
  • Add to Cart