COM448686172 Combi-Blocks, Inc.

methyl (2S)-2-amino-4-(ethylcarbamoyl)butanoate

  • MDL number MFCD27578275
  • Cas Number : 1000788-95-3
  • Molecular Weight : 188.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0653

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0653
Availability details for
Quantity
  • Availability
  • Add to Cart