COM448686174 Combi-Blocks, Inc.

methyl 3-[(triphenylmethyl)sulfanyl]propanoate

  • MDL number MFCD27578276
  • Cas Number : 500309-29-5
  • Molecular Weight : 362.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0656

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0656
Availability details for
Quantity
  • Availability
  • Add to Cart