COM448686180 Combi-Blocks, Inc.

Methyl 2-n-propyloxybenzoate

  • MDL number MFCD07780728
  • Cas Number : 18167-33-4
  • Molecular Weight : 194.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0663

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0663
Availability details for
Quantity
  • Availability
  • Add to Cart