COM448686185 Combi-Blocks, Inc.

1-tert-butyl 4-methyl (2S)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}butanedioate

  • MDL number MFCD27578279
  • Cas Number : 858034-72-7
  • Molecular Weight : 425.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0670

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0670
Availability details for
Quantity
  • Availability
  • Add to Cart