COM448686192 Combi-Blocks, Inc.

methyl (2R)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl](methyl)amino}-4-methylpentanoate

  • MDL number MFCD27578283
  • Cas Number : 1820575-65-2
  • Molecular Weight : 381.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0678

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0678
Availability details for
Quantity
  • Availability
  • Add to Cart