COM448686193 Combi-Blocks, Inc.

Methyl (2r)-2-[(tert-butoxy)carbonylamino]-3-(4-pyridyl)propanoate

  • MDL number MFCD11978148
  • Cas Number : 1241676-69-6
  • Molecular Weight : 280.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0680

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0680
Availability details for
Quantity
  • Availability
  • Add to Cart