COM448686196 Combi-Blocks, Inc.

Methyl (2r)-2-amino-3-(2-chlorophenyl)propanoate

  • MDL number MFCD08058197
  • Cas Number : 103616-88-2
  • Molecular Weight : 213.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0683

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0683
Availability details for
Quantity
  • Availability
  • Add to Cart