COM448686197 Combi-Blocks, Inc.

methyl 1-(3,4-difluorophenyl)-5-oxopyrrolidine-3-carboxylate

  • MDL number MFCD27578284
  • Cas Number : 1820604-85-0
  • Molecular Weight : 255.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0685

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0685
Availability details for
Quantity
  • Availability
  • Add to Cart