COM448686199 Combi-Blocks, Inc.

Methyl (2r)-2-aminopentanoate

  • MDL number MFCD19204072
  • Cas Number : 21685-17-6
  • Molecular Weight : 131.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0691

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0691
Availability details for
Quantity
  • Availability
  • Add to Cart