COM448686202 Combi-Blocks, Inc.

2-tert-butyl 3-methyl (3S)-3,4-dihydro-1H-isoquinoline-2,3-dicarboxylate

  • MDL number MFCD27578287
  • Cas Number : 252008-94-9
  • Molecular Weight : 291.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0696

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0696
Availability details for
Quantity
  • Availability
  • Add to Cart