COM448686206 Combi-Blocks, Inc.

[1,1'-Biphenyl]-4-acetic acid methyl ester

  • MDL number MFCD02056549
  • Cas Number : 59793-29-2
  • Molecular Weight : 226.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0702

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0702
Availability details for
Quantity
  • Availability
  • Add to Cart