COM448686207 Combi-Blocks, Inc.

Methyl trans-4-propylcyclohexanecarboxylate

  • MDL number MFCD27981349
  • Cas Number : 117169-72-9
  • Molecular Weight : 184.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0704

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0704
Availability details for
Quantity
  • Availability
  • Add to Cart