COM448686218 Combi-Blocks, Inc.

Dimethyl 5-methylisophthalate

  • MDL number MFCD00082721
  • Cas Number : 17649-58-0
  • Molecular Weight : 208.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0724

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0724
Availability details for
Quantity
  • Availability
  • Add to Cart