COM448686224 Combi-Blocks, Inc.

Methyl 2-(4-bromophenyl)thiazole-4-carboxylate

  • MDL number MFCD15142858
  • Cas Number : 1208081-39-3
  • Molecular Weight : 298.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 97
Storage temp. Refrigerated

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0730
Availability details for
Quantity
  • Availability
  • Add to Cart