COM448686225 Combi-Blocks, Inc.

Methyl 2-[(4-chlorophenyl)methyl]-1,3-thiazole-4-carboxylate

  • MDL number MFCD23147784
  • Cas Number : 154220-01-6
  • Molecular Weight : 267.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0731

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0731
Availability details for
Quantity
  • Availability
  • Add to Cart