COM448686230 Combi-Blocks, Inc.

Methyl 2-([(benzyloxy)carbonyl](methyl)amino)acetate

  • MDL number MFCD12563551
  • Cas Number : 100391-80-8
  • Molecular Weight : 237.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0740

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0740
Availability details for
Quantity
  • Availability
  • Add to Cart