COM448686231 Combi-Blocks, Inc.

Methyl (2s)-2-phenyl-2-[(phenylmethoxy)carbonylamino]acetate

  • MDL number MFCD15478864
  • Cas Number : 147780-61-8
  • Molecular Weight : 299.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0743

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0743
Availability details for
Quantity
  • Availability
  • Add to Cart