COM448686234 Combi-Blocks, Inc.

methyl (2R,3R,4R,5R)-2,3,5,6-tetrahydroxy-4-{[(2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}hexanoate

  • MDL number MFCD27578294
  • Cas Number : 121436-08-6
  • Molecular Weight : 372.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0748

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0748
Availability details for
Quantity
  • Availability
  • Add to Cart