COM448686235 Combi-Blocks, Inc.

methyl 2-[(4-hydroxyphenyl)carbonyl]benzoate

  • MDL number MFCD27578295
  • Cas Number : 21147-23-9
  • Molecular Weight : 256.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0749

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0749
Availability details for
Quantity
  • Availability
  • Add to Cart