COM448686237 Combi-Blocks, Inc.

Methyl (2R)-2-{[(benzyloxy)carbonyl]amino}-4-methylpentanoate

  • MDL number MFCD25974201
  • Cas Number : 73397-22-5
  • Molecular Weight : 279.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0754

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0754
Availability details for
Quantity
  • Availability
  • Add to Cart