COM448686238 Combi-Blocks, Inc.

(S)-Methyl 2-amino-2-(4-fluorophenyl)acetate

  • MDL number MFCD08704819
  • Cas Number : 170902-74-6
  • Molecular Weight : 183.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 98

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0755
Availability details for
Quantity
  • Availability
  • Add to Cart