COM448686240 Combi-Blocks, Inc.

2,5-Difluoro-4-methylbenzoic acid methyl ester

  • MDL number MFCD16250137
  • Cas Number : 1355488-71-9
  • Molecular Weight : 186.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0757

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0757
Availability details for
Quantity
  • Availability
  • Add to Cart