COM448686242 Combi-Blocks, Inc.

methyl 5-bromo-2-acetamidopyridine-4-carboxylate

  • MDL number MFCD27578296
  • Cas Number : 1820684-64-7
  • Molecular Weight : 273.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0760

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0760
Availability details for
Quantity
  • Availability
  • Add to Cart